missing translation for 'onlineSavingsMsg'
Learn More

pH Buffer Pouches, Oakton™

Product Code. p-2454309
Click to view available options
Quantity:
20 Ea.
pH:
4.01
10.00
Unit Size:
Pack of 20
This item is not returnable. View return policy

Product Code. 17967242

Brand: Oakton 3565301

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Oakton™ pH buffer pouches; package of 20 Economical pouches are accurate and convenient.±0.01 accuracy at 77°°F (25°°C)

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4.01
Physical Form Liquid
Quantity 20 Ea.
Solution Type pH Buffer
Chemical Name or Material Oakton pH buffer pouches
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.