Learn More
o-Tolylacetic acid, 99%, Thermo Scientific Chemicals
Catalog Number 11422448
Click to view available options
Quantity:
5 g
25 g
100 g
644-36-0
C9H10O2
150.177
MFCD00004328
RZWGTXHSYZGXKF-UHFFFAOYSA-N
o-tolylacetic acid, 2-methylphenylacetic acid, 2-methylphenyl acetic acid, 2-2-methylphenyl acetic acid, 2-o-tolyl acetic acid, 2-tolylacetic acid, 2-o-tolylacetic acid, o-methylphenylacetic acid, o-tolyacetic acid
69519
2-(2-methylphenyl)acetic acid
CC1=CC=CC=C1CC(=O)O
644-36-0
C9H10O2
150.177
MFCD00004328
RZWGTXHSYZGXKF-UHFFFAOYSA-N
o-tolylacetic acid, 2-methylphenylacetic acid, 2-methylphenyl acetic acid, 2-2-methylphenyl acetic acid, 2-o-tolyl acetic acid, 2-tolylacetic acid, 2-o-tolylacetic acid, o-methylphenylacetic acid, o-tolyacetic acid
69519
2-(2-methylphenyl)acetic acid
CC1=CC=CC=C1CC(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 86°C to 90°C
Quantity 5 g
Beilstein 1936952
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material o-Tolylacetic acid