Learn More
Thermo Scientific O-Rings, Viton
Product Code. 15894907
Change view
Click to view available options
For Use With (Equipment):
1000 mL Bottle
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15894907 1000 mL Bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15894907 Supplier Thermo Scientific Supplier No. 75003020
  • ONLINE EXCLUSIVE PRICE
    505.00 NOK
    452.74 NOK / Pack of 6
    Save 52.26 NOK
    10% Off
In Stock
Replacement O-Rings, Viton, for 1000 mL bottles.
  • Replacement O-Rings x 6, Viton, for 1000 mL bottles
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 1000 mL Bottle
Disposable Reusable
Quantity 6 Pk.
Product Type O-ring