Learn More
O-Methyl-L-serine hydrochloride, 95%
Product Code. 15489307
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15489307 250 mg 250mg
15479307 1 g 1g
2 options
Product Code. 15489307
Brand: Thermo Scientific Alfa Aesar H52562.MD
Request Bulk or Custom Format
CAS: 336100-47-1 | C4H10ClNO3 | 155.58 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 336100-47-1
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.58
MDL Number MFCD03840366
InChI Key NLDVBPFPOCWZCE-UHFFFAOYNA-N
Synonym s-2-amino-3-methoxypropanoic acid hydrochloride, s-2-amino-3-methoxy-propionic acid hydrochloride, s-2-amino-3-methoxypropionic acid hydrochloride, o-methyl-l-serine hydrochloride, 2s-2-amino-3-methoxypropanoic acid hydrochloride, l-serine, o-methyl-, hydrochloride 1:1, l-o-methylserine hydrochloride, l-serine, o-methyl-, hydrochloride, s-2-amino-3-methoxy-propionicacidhydrochloride, s-2-amino-3-methoxypropanoic acid hcl
PubChem CID 16038806
IUPAC Name 2-amino-3-methoxypropanoic acid hydrochloride
SMILES Cl.COCC(N)C(O)=O
Specifications
Melting Point 182°C to 183°C
Quantity 250 mg
Formula Weight 155.58
Percent Purity 97%
Chemical Name or Material O-Methyl-L-serine hydrochloride