Learn More
o-Flutamide, TRC
Product Code. 30135785
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.50mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135785 50 mg 50mg
30137007 2.5 mg 2.50mg
30005801 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135785 Supplier Toronto Research Chemicals Supplier No. TRCF59885550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 151262-93-0
Molecular Formula C11 H11 F3 N2 O3
Molecular Weight (g/mol) 276.21
Synonym 2-Methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide, Flutamide Imp. F (EP)
IUPAC Name 2-methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide
SMILES CC(C)C(=O)Nc1cc(ccc1[N+](=O)[O-])C(F)(F)F
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C11H11F3N2O3/c1-6(2)10(17)15-8-5-7(11(12,13)14)3-4-9(8)16(18)19/h3-6H,1-2H3,(H,15,17)
Formula Weight 276.0722
Chemical Name or Material 2-Methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide