Learn More
o-Aminoazotoluene, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 4-Amino-2,3-dimethylazobenzene |
| CAS | 97-56-3 |
| Molecular Formula | C14 H15 N3 |
| Quantity | 25 g |
| Synonym | Benzenamine, 2-methyl-4-[(2-methylphenyl)azo]- (9CI), C.I. Solvent Yellow 3 (8CI), o-Toluidine, 4-o-tolylazo- (6CI), 2-Methyl-4-[2-(2-methylphenyl)diazenyl]benzenamine, 2,3'-Dimethyl-4'-aminoazobenzene, 2-Aminoazotoluene, 2-Methyl-4-[(2-methylphenyl)diazenyl]amine, 2-Methyl-4-[(o-tolyl)azo]aniline, 2',3-Dimethyl-4-aminoazobenzene, 4-(o-Tolylazo)-o-toluidine, 4-Amino-2',3-dimethylazobenzene, Brasilazina Oil Yellow R, C.I. 11160, C.I. 11160B, Fast Garnet GBC Base, Fast Yellow AT, Fat Yellow B, Hidaco Oil Yellow, NSC 1797, NSC 26821, OAAT, Oil Yellow 21, Oil Yellow 2681, Oil Yellow 2R, Oil Yellow AT, Oil Yellow C, Oil Yellow I, Organol Yellow 2T, Solvent Yellow 3, Somalia Yellow R, o-AT, o-Aminoazotoluene |
| InChI Formula | InChI=1S/C14H15N3/c1-10-5-3-4-6-14(10)17-16-12-7-8-13(15)11(2)9-12/h3-9H,15H2,1-2H3 |
| SMILES | Cc1ccccc1N=Nc2ccc(N)c(C)c2 |
| IUPAC Name | 2-methyl-4-[(E)-(2-methylphenyl)diazenyl]aniline |
| Molecular Weight (g/mol) | 225.29 |
| Formula Weight | 225.1266 |