Learn More
Cole-Parmer™ Nylon Turbine Flowmeter
Specifications
Specifications
| Type | Turbine Flowmeter |
| Accuracy | ±5% (Reading) |
| Float Material | Nylon |
| Pressure Rating | 150 psig |
| Connections | 1 in. NPT (F) |
| Flow Rate Range | 1 to 10 LPM |
| For Use With (Application) | Low-cost solution to light-duty process applications |
| Height (Metric) | 64 mm |
| Power Supply Details | Two 3 V Lithium Batteries |
| Repeatability | ±0.1% |
| Show More |