Learn More
Brady™ Nylon Ties
Product Code. p-8006765
missing translation for 'orderingAttributeHoverText'
Color:
Blue
Orange
Purple
Red
White
Yellow
missing translation for 'unitSize'
1 pack
Product Code. 11776885
missing translation for 'mfr': Brady™ 229955
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
  • Cross-laminated polyethylene
  • Matte finish
Specifications
Type Ties
Material Nylon
Color White
Length (Metric) 203.00 mm
Width (Metric) 4 mm
Dimensions (L x W) 203 x 3.60 mm
Quantity 100/Pk.
For Use With (Application) Ideal for color-coding hard to mark items