Learn More
Masterflex™ Nylon Straight Tubing Connector
Product Code. 12314618
Change view
Click to view available options
For Use With:
14.6mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
12314618 14.6mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12314618 Supplier Masterflex™ Supplier No. 3061210
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Diameter (Metric) Inner 14.6 mm
Material Nylon
For Use With 14.6mm
Min. Operating Temperature (Metric) -46°C
Max. Operating Temperature (Metric) 121°C
Product Type Straight Tubing Connector