Learn More
Masterflex™ Nylon Straight Barbed Union
Product Code. 12334618
Click to view available options
For Use With:
9.52mm
12.7mm
19.05mm
25.4mm
Unit Size:
Pack of 10
Product Code. 12334618
Brand: Masterflex™ 3061218
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Diameter (Metric) Inner 9.52 mm
Material Nylon
For Use With 9.52mm
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 10 Pack
Max. Operating Temperature (Metric) 121°C
Product Type Straight Barbed Union