Learn More
Ansell Edmont™ Marigold™ Picolon Confort™ Polyamide Gloves
Product Code. p-8010803
Change view
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
1 pair
12 pairs
Each
Product Code. Size unitSize
12961135 X-Large 1 pair
15962102 Large 12 pairs
12382209 Medium Each
3 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12961135
Brand: Ansell Edmont™ C357100
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Providing good resistance to abrasion and small blade cut hazards. Ansell Edmont™ Marigold™ Picolon Confort™ Polyamide Gloves are ideal for the handling and assembly of slightly sharp and slippery objects.
  • Dots on palm and fingers for better grip
  • Textile liner made of high elasticity polyamide thread
  • Fine, stretchable and comfortable stitches
  • Knitted with a seamless non-stop process
  • Lint-free
TRUSTED_SUSTAINABILITY
Specifications
Construction Seamless Knit
Coating Material PVC
Color White with Blue Dots
Size X-Large
Glove Cuff Style Knit
Liner Material Polyamide
Resistant To Abrasion and small cuts
Length (Metric) 260 mm