Learn More
Masterflex™ Nylon Female Luer Fitting
Click to view available options
For Use With:
Female Luer x 1/8 in. Wide Bore Hose Barb
Female Luer x 1/8-27 NPT Elbow
Female Luer x 3/32 in. Hose Barb
Female Luer x 3/32 in. Wide Bore Hose Barb
Unit Size:
Pack of 25
Description
- Standard hose barb for use with flexible tubing
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination
- Wide-bore hose barb for enhanced fluid path flow
Specifications
Specifications
| Material | Nylon |
| For Use With | Female Luer x 1/8 in. Wide Bore Hose Barb |
| Packaging Quantity | 25 Pack |
| Product Type | Female Luer Fitting |