Learn More
Masterflex™ Nylon Clamps for Sanitary Fittings
Description
Maintain sanitary conditions with these clamps. Cole-Parmer™ Nylon Clamp for Sanitary Fittings feature excellent chemical resistance and low leachable characteristics.
- Low leachable characteristics
- Sterilize with Ethylene Oxide (ETO)
- Compatible with Tri-Clamp™ fittings
Specifications
Specifications
| Material | Nylon |
| Packaging Quantity | 1 Ea. |
| Width (Metric) | 25.4 to 38.1 mm |
| Product Type | Tubing Clamp |