Learn More
B2M™ Nylon Nail Brush
Product Code. 11738602
Change view
Click to view available options
Color:
White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11738602 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11738602 Supplier B2M™ Supplier No. 40294
  • ONLINE EXCLUSIVE PRICE
    282.23 NOK
    78.85 NOK / Pack of 1
    Save 203.38 NOK
    72% Off
Estimated Shipment: 15-12-2026
Designed to clean nails, hands and forearms. B2M™ Nylon Nail Brush has hard nylon bristles to remove debris and microorganisms in order to prevent the spread of biological contaminants and infectious disease. It can be autoclaved for effective sterilization.
  • Thick handle fits easily in the palm
  • Flat brush with highly resistant nylon bristles
  • Can be autoclaved for use in sterile environments
Specifications
Product Type Nail Brush
Autoclavable Autoclavable
Color White
For Use With (Application) Cleaning of nails, hands and forearms
Quantity 1 Ea.