Learn More
Masterflex™ Nylon Barbed Fitting
Product Code. 12487250
Click to view available options
For Use With:
12.7mm
15.8mm
19.05mm
3.17mm
6.35 x 6.35mm
9.52 x 6.35mm
Unit Size:
Pack of 10
Product Code. 12487250
Brand: Masterflex™ 3061526
Only null left
Specifications
Diameter (Metric) Inner 15.8 mm
Material Nylon
For Use With 15.8mm
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 10 Pack
Thread Size 0.5 in. NPT
Max. Operating Temperature (Metric) 121°C
Product Type Barbed Fitting