Learn More
Masterflex™ Nylon Barbed Fitting
Specifications
Specifications
| Material | Nylon |
| For Use With | 6.35 x 6.35mm |
| Min. Operating Temperature (Metric) | -46°C |
| Packaging Quantity | 10 Pack |
| Thread Size | 0.25 in. NPT |
| Max. Operating Temperature (Metric) | 121°C |
| Product Type | Barbed Fitting |