Learn More
Masterflex™ Nylon Barbed Fitting Reducing Union
Product Code. 12457260
Click to view available options
For Use With:
115.87 x 12.7mm
12.7 x 12.7mm
19.05 x 12.7mm
6.35 x 3.175mm
Unit Size:
Pack of 10
Product Code. 12457260
Brand: Masterflex™ 3061626
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material Nylon
For Use With 115.87 x 12.7mm
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 10 Pack
Max. Operating Temperature (Metric) 121°C
Product Type Barbed Fitting Reducing Union