Learn More
NVP-CGM097, MedChemExpress
Product Code. 16467006
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Product Code. 16467006
Brand: Medchem Express HY15954
MedChemExpress NVP-CGM097 is a potent and selective MDM2 inhibitor with IC50 of 1.7±0.1 nM for hMDM2.
NVP-CGM097 is a potent and selective MDM2 inhibitor with IC50 of 1.7±0.1 nM for hMDM2.
Specifications
Chemical Name or Material NVP-CGM097
CAS 1313363-54-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C38H47ClN4O4
Quantity 100 mg
Synonym CGM097
Solubility Information DMSO : ≥ 50 mg/mL (75.84 mM)
SMILES ClC(C=C1)=CC=C1[C@@H](C2=CC(OC(C)C)=C(OC)C=C2C3)N(C4=CC=C(N(C[C@@H]5CC[C@@H](N6CC(N(C)CC6)=O)CC5)C)C=C4)C3=O
Molecular Weight (g/mol) 659.26
Formula Weight 659.26
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.