missing translation for 'onlineSavingsMsg'
Learn More

NUP54 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16855794 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16855794 20 μL 20µL
16845794 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16855794

Brand: Proteintech 162321AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. This gene encodes a member of the phe-gly repeat-containing nucleoporin subset.

Specifications

Antigen NUP54
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NUP54
Gene Accession No. P70582, Q7Z3B4, Q8BTS4
Gene Alias 54 kDa nucleoporin, nucleoporin 54kDa, Nucleoporin p54, NUP54
Gene Symbols Nup54
Host Species Rabbit
Immunogen NUP54 Fusion Protein Ag8882
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269113, 53371, 53372
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.