missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP50 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20798-1-AP-20UL
This item is not returnable.
View return policy
Description
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. NUP50 is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. NUP50 may serve as a binding site on the nuclear side of the pore complex for export receptor-cargo complexes. It interacts with multiple transport receptor proteins including p27Kip1. This interaction is required for correct intracellular transport and degradation of p27Kip1.Specifications
| NUP50 | |
| Polyclonal | |
| Unconjugated | |
| NUP50 | |
| 50 kDa nucleoporin, nucleoporin 50kDa, Nucleoporin Nup50, NUP50 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10762, 18141, 25497 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08587, Q9JIH2, Q9UKX7 | |
| Nup50 | |
| NUP50 Fusion Protein Ag14710 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction