missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP153 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14189-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Nuclear pore complexes regulate the transport of macromolecules between the nucleus and cytoplasm. They are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. Nucleoporins are glycoproteins found in nuclear pores and contain characteristic pentapeptide XFXFG repeats as well as O-linked N-acetylglucosamine residues oriented towards the cytoplasm. The protein encoded by this gene has three distinct domains: a N-terminal region containing a pore targeting and an RNA-binding domain , a central region containing multiple zinc finger motifs, and a C-terminal region containing multiple XFXFG repeats. Alternative splicing results in multiple transcript variants of this gene.Specifications
NUP153 | |
Polyclonal | |
Unconjugated | |
NUP153 | |
153 kDa nucleoporin, HNUP153, N153, nucleoporin 153kDa, Nucleoporin Nup153, NUP153 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
218210, 25281, 9972 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49790, P49791 | |
NUP153 | |
NUP153 Fusion Protein Ag5519 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |