Learn More
Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes
GREENER_CHOICE
Product Code. 12617316
Change view
Click to view available options
Holds:
1 to 2 mL Cryogenic Tubes
2 mL Cryogenic Tubes
3.6 to 5 mL Cryogenic Tubes
5 mL Cryogenic Tubes
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
12617316 1 to 2 mL Cryogenic Tubes Pack of 10
12586528 2 mL Cryogenic Tubes Pack of 10
12596528 5 mL Cryogenic Tubes Pack of 10
12556528 3.6 to 5 mL Cryogenic Tubes Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12617316 Supplier Thermo Scientific™ Supplier No. 331827
Store tubes for automated picking/placing, liquid handling and other high throughput applications using Nunc Universal Latch Rack for Cryogenic Tubes.
Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes. Features a transparent cover with options for tubes of several sizes.
  • Non-sterile
Ordering Information:
Case of 10
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
For Use With (Equipment) 1-2mL Standard Cryotubes, non-sterile
Holds 1 to 2 mL Cryogenic Tubes
Quantity 10/pk., 10/cs.
Type Cryogenic Tube Storage Rack (w/Latch)