Learn More
Thermo Scientific™ Nunc™ Screwdrivers for Cryobank and Bank-It™ Vial Systems, manual
Product Code. p-4520570
Change view
Click to view available options
Type:
Electric Screwdriver
Manual Screwdriver
Plug Type:
European
Not Applicable
Unit Size:
Each
Product Code. Type Plug Type unitSize
10709153 Manual Screwdriver Not Applicable Each
10401633 Electric Screwdriver European Each
2 options
Product Code. 10709153
Brand: Thermo Scientific™ 369643
Only null left
Add to basket
Add to basket
Quickly and easily cap and decap Thermo Scientific™ Nunc™ Cryobank tubes using these screwdrivers, available in manual and electric models to suit your application.
Quickly and easily cap and decap Nunc Cryobank tubes using Thermo Scientific™ Nunc™ Screwdrivers for Cryobank and Bank-It™ Systems, available in manual and electric models to suit your application. Nunc Cryobank vials provide the customer with superior tracking of their cryovials using a 2D code, the highest storage density on the market and the ability to automate the capping and decapping of the vials. The tubes click into the rack and can be handled without the risk of accidentally dropping a vial.
Screwdriver socket retains the vial cap, allowing one-handed manipulation of the cap.
TRUSTED_SUSTAINABILITY
Specifications
Description Manual Screwdriver
Type Manual Screwdriver
Coding 2D Barcode
Vial Type Cryobank, Bank-It
Color Red, Black
Plug Type Not Applicable
For Use With (Equipment) Cryobank and Bank-It Systems