Learn More
Thermo Scientific™ Nunc™ Press Out Tool for Cryobank and Bank-It™ Vial Systems
Product Code. 10753754
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10753754 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10753754 Supplier Thermo Scientific™ Supplier No. 374009
  • ONLINE EXCLUSIVE PRICE
    24230.00 NOK
    22215.98 NOK / Pack of 1
    Save 2014.02 NOK
    8% Off
View all promotions
 Restricted Item
In Stock
Minimize freeze/thaw by using the Thermo Scienctific™ Nunc™ Press Out Tool for Cryobank and Bank-It™ Systems, which allows for rapid retrieval of Cryobank vials.
Locate and remove a desired tube quickly and easily, without allowing frozen samples to thaw by using the Thermo Scientific™ Nunc™ Press Out Tool for Cryobank and Bank-It™ Vial Systems.
Features:
  • Selected vials are released by levers and can be easily harvested
  • Selected position can be chosen based on data base output
  • Pegs can be placed and checked while the sample/neighboring samples remain cold
  • Cut-off corner ensures the rack is orientated correctly
TRUSTED_SUSTAINABILITY
Specifications
Description Nunc Cryobank and Bank-it Rack, Tube Press out Tool, 1 Each
Vial Type Cryobank
Color Silver
For Use With (Equipment) Cryobank and Bank-It Systems
Quantity 1 Ea.
Type Tube Press Out Tool