Learn More
Thermo Scientific™ Nunc™ Latch Rack for 1.8 mL Externally-Threaded Universal Tubes Product Code.: 13426123
Only undefined left
Add to basket
Thermo Scientific™ Nunc™ Latch Rack for 1.8 mL Externally-Threaded Universal Tubes
GREENER_CHOICE
Product Code. 13426123
None, Pack of 10
Click to view available options
Barcode:
2D, linear and human readable
None
Unit Size:
Pack of 10
Product Code. 13426123
Brand: Thermo Scientific™ 331830
Only undefined left
Add to basket
Only undefined left
Add to basket
Versatile tubes for use inside a large storage system, or in smaller scale for traditional cryostorage in dewars.
Use versatile Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes or non-coded.
Scalable
  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10x10 arrays
  • External threads limit contamination
  • Automation-pickable in Universal Latch Rack
Innovative Universal Cap Design
  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
  • Available with a permanently-bonded, unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
  • 374502NOV is featured with 1D (linear) barcode and human readable on the side that matches 2D barcode on the bottom of the tube
Stringent Quality Control
  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 106 and is made with class VI medical grade resins
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY
Specifications
Barcode None
Capacity (Metric) 1.8 mL (Tube)
Description Nunc latch rack
Color Beige
Type Latch Rack
For Use With (Equipment) Cryobank
Holds Externally-Threaded Universal Tubes
Includes 10 empty racks
Quantity 1/Pk., 10/Cs.
Sterility Non-sterile
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ Latch Rack for 1.8 mL Externally-Threaded Universal Tubes >