Learn More
Thermo Scientific™ Nunc™ Disposable Loops and Needles
Product Code. 10659971
Change view
Click to view available options
Volume:
1 μL
10 μL
Quantity:
Carton of 1000
Carton of 600
Unit Size:
Pack of 1000
Pack of 2400
Pack of 4000
Pack of 600
Product Code. Volume Quantity unitSize
10659971 1 μL Carton of 600 Pack of 2400
10048750 10 μL Carton of 1000 Pack of 1000
11391575 10 μL Carton of 1000 Pack of 4000
10344741 1 μL Carton of 1000 Pack of 1000
11301585 1 μL Carton of 1000 Pack of 4000
12687093 1 μL Carton of 600 Pack of 600
12780006 10 μL Carton of 600 Pack of 600
11351585 10 μL Carton of 600 Pack of 2400
8 options
Product Code. 10659971
Brand: Thermo Scientific™ 254410
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1 μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Volume 1 μL
Material Polystyrene
Sterility Sterile
Color Clear
Product Type Inoculating Loop (Clear)
Quantity Carton of 600