Learn More
Thermo Scientific™ Nunc™ Coded Cryobank Vial Systems
GREENER_CHOICE
Product Code. 12713386
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
0.5 mL
1 mL
2 mL
5 mL
Closure Color:
Blue
Clear
Green
Red
Coding:
2D Barcode
None
Packaging:
10 racks of 48/case
192/cs
Black low rack, 96/960
Bulk, 11 bags of 48 tubes/case
Bulk, 20 bags of 48 tubes/case
Locking latch rack with lid, 48/480
missing translation for 'unitSize'
Pack of 10
Pack of 192
Pack of 480
Pack of 528
Pack of 96
Pack of 960
Product Code. 12713386
missing translation for 'mfr': Thermo Scientific™ 374221LOCK
Only null left
Add to basket
Add to basket
Take cryostorage to the next level with this vial system featuring 2D barcode tubes in standard microplate format racks for benchtop use or automated applications.
Take traditional cryostorage to the next level while accommodating denser storage capability compared to standard cryoboxes. Thermo Scientific™ Nunc™ Coded Cryobank Vial Systems feature laser-etched 2D barcodes and standard microplate format racks. Compatible with the complete line of 2D barcode readers, capping/decapping equipment and labeling options, Cryobank tubes can be used in manual or automated storage of biological samples down to vapor phase liquid nitrogen. Cryobank products meet leading certification requirements and are available in a volume range of 0.5 to 5.0mL.
Secure Tracking
  • Permanently attached, laser etched 2D barcode on the bottom of the tube allows you to scan and decode the entire rack at once without removing them from the rack, and allows data to be associated with each tube
  • Solid one piece construction eliminates the risk of labels falling off during cryogenic storage, preserving the identity of valuable sample material
  • Range of 2D reading products to decode individual tubes or complete microplate footprint racks. Readers allow flexible output to store data in multiple database formats
Superior Storage Format
  • Cryobank tubes are available in space conscious microplate format racks or can be stored in traditional cryoboxes, accommodating your preferred storage format
  • Microplate format is compatible with your existing multichannel manual pipettes and automated liquid handling equipment
  • Unique anti-rotation feature allows compatibility with automated decapping/ capping equipment in single, 8-channel, or full rack options to accommodate multiple throughput requirements
  • TPE gasketed polypropylene cap provides superior seal during high or low temperature storage
  • Low binding surface allows maximum recovery of low concentration samples
Certified to meet your storage needs
  • Certified DNAse/RNAse free (0.5/1.0mL), non-pyrongenic (LAL-test), non-toxic (USP class VI test)
  • Sterility assurance level 10-6 for the most sensitive sample storage
Stringent Quality Control
  • 2D barcodes are scanned for readability and checked against a full database of previous barcodes to ensure no duplicates across the Cryobank storage tube product line
  • Leak testing validation performed during the manufacturing process to ensure sample integrity is maintained
TRUSTED_SUSTAINABILITY
Specifications
Description 2 ml Cryobank 2D code racked Sterile
Closure Color Clear
Capacity (Metric) 2 mL
Sterility Sterile
Type Coded Cryobank Vial System
Coding 2D Barcode
Vial Type Cryobank
Color Clear (Closure)
Packaging 10 racks of 48/case