Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System
Product Code. 13101594
Change view
Click to view available options
Tray Layers:
1
2
4
10
40
Culture Area:
1264 cm²
25,280 cm²
2528 cm²
632 cm²
6320 cm²
No. per Case:
10/Cs.
2/Cs.
6/Cs.
8/Cs.
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 5
Pack of 6
Pack of 8
Product Code. Tray Layers Culture Area No. per Case unitSize
13101594 2 1264 cm² 6/Cs. Pack of 5
10344321 40 25,280 cm² 2/Cs. Pack of 2
12677093 4 2528 cm² 10/Cs. Each
10113922 4 2528 cm² 10/Cs. Pack of 10
10049040 10 6320 cm² 2/Cs. Pack of 2
10634551 1 632 cm² 8/Cs. Pack of 8
10394221 2 1264 cm² 6/Cs. Pack of 6
10604901 10 6320 cm² 6/Cs. Pack of 6
8 options
Product Code. 13101594
Brand: Thermo Scientific™ 167695
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems.
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.
  • Ideal for adherent cells
  • Boost production efficiency with more surface area in a small footprint
  • Reduce contamination risks with a closed system
Warranty: 90 days
TRUSTED_SUSTAINABILITY
Specifications
Description Original Cell Factory
Material Polystyrene
No. per Case 6/Cs.
No. per Pack 1 Ea.
Product Type Cell Factory System
Surface Coating Nunclon™ Delta
Green Features Less waste, renewable material
Closure Type Adapter Cap and Port Cover Cap
Culture Area 1264 cm²
Surface Treatment Nunclon Delta
Tray Layers 2
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.