Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System Accessories
Product Code. 11756745
Change view
Click to view available options
Product Type:
Adapter Closure
Airvent Filter
Airvent Filter, 0.22 μm
Cover Cap
FEP Connector
Filter Adapter Cap, Sterile
Filter Assembly, 200 cm⊃2
Filter Assembly, 37 mm
HDPE Connector
PC Connector
System Start-Up Kit
Tubing Set, Female MPC
Tubing Set, Male MPC
Vent Cap
Vent/Close Cap
For Use With (Equipment):
Cell Factory Systems (All)
EasyFill Cell Factory Systems
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 12
Pack of 2
Pack of 20
Pack of 40
Product Code. Product Type For Use With (Equipment) unitSize
11756745 Airvent Filter Cell Factory Systems (All) Pack of 10
10562944 PC Connector Cell Factory Systems (All) Pack of 10
10330525 Vent Cap EasyFill Cell Factory Systems Pack of 10
10377873 Filter Adapter Cap, Sterile Cell Factory Systems (All) Pack of 20
12663096 Cover Cap Cell Factory Systems (All) Pack of 40
10144302 Airvent Filter, 0.22 μm Cell Factory Systems (All) Pack of 2
11306891 Filter Assembly, 200 cm⊃2 EasyFill Cell Factory Systems Pack of 2
11376881 Filter Assembly, 37 mm EasyFill Cell Factory Systems Pack of 2
10092151 Vent/Close Cap Cell Factory Systems (All) Pack of 100
15390332 Tubing Set, Female MPC Cell Factory Systems (All) Pack of 2
15390292 Tubing Set, Male MPC Cell Factory Systems (All) Each
15778032 Tubing Set, Male MPC Cell Factory Systems (All) Pack of 2
11377734 HDPE Connector Cell Factory Systems (All) Pack of 10
15357026 FEP Connector Cell Factory Systems (All) Pack of 2
12823133 FEP Connector Cell Factory Systems (All) Pack of 2
11366881 Adapter Closure EasyFill Cell Factory Systems Pack of 12
10274451 System Start-Up Kit Cell Factory Systems (All) Each
17 options
Product Code. 11756745
Brand: Thermo Scientific™ 179553
Only null left
Add to basket
Add to basket
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Connections Made Easy:
  • Plug a media bag or tubing set to any Cell Factory system for enhanced aseptic connectivity for filling and harvesting
Additional Features:
  • Save time and reduce costs
  • Minimize cleaning processes such as autoclaving and associated validations
  • Prevent build up of back pressure during filling application
  • Get additional air venting with the venting options
TRUSTED_SUSTAINABILITY
Specifications
Description Airvent Filter
For Use With (Equipment) Cell Factory Systems (All)
No. per Case 10
No. per Pack 1
Product Type Airvent Filter
Sterility Sterile
Bottom Shape Flat
Pore Size (Metric) 1µm
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.