Learn More
Thermo Scientific™ Nunc™ 1.8mL Externally-Threaded Universal Tubes with 1D (linear) code on the side and 2D code on tube bottom, not racked
Use this versatile tube either inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars.
Marca: Thermo Scientific™ 374502NOV
Descrizione
Use versatile Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes either inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes or non-coded.
Scalable
- Tubes and rack are ideal for both lower-throughput and high-throughput use
- Feature graduations and white patch for manual marking
- Can be used in traditional cyrostorage boxes made for 10x10 arrays
- External threads limit contamination
- Automation-pickable in Universal Latch Rack
Innovative Universal Cap Design
- Single-piece construction with molded-in gasket
- Cannot be over-tightened manually or with automated capping tools
- Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
- Permanently-bonded,unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
- 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
- Scan the tube to access sample information and identify the contents
Stringent Quality Control
- Every 2D barcoded storage tube is scanned to ensure readability
- Barcodes are checked against database of previously-assigned codes to prevent duplicates
- Every tube has a Sterility Assurance Level of 10superscript 6 and is made with class VI medical grade resins
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.