Learn More
Macherey-Nagel™ NucleoVac Vacuum Regulator
Beskrivelse
Vacuum filtration columns are used to isolate and purify nucleic acids, including plasmid DNA. A reliable vacuum source is critical for successful manual processing. A vacuum regulator is recommended to control the vacuum pressure levels, which impacts the flow rate of the sample through the column. In addition, a regulator controls the vacuum time to ensure complete filtration of the sample.
The NucleoVac Vacuum Regulator is designed for use with the NucleoVac Vacuum Manifold and with various DNA isolation/purification column systems, including NucleoSpin™, NucleoFast™ and NucleoSnap kits.
The NucleoVac Vacuum Regulator is designed for use with the NucleoVac Vacuum Manifold and with various DNA isolation/purification column systems, including NucleoSpin™, NucleoFast™ and NucleoSnap kits.
Tekniske data
Tekniske data
| Product Type | NucleoVac Vacuum Regulator |
| Description | For use with NucleoVac Vacuum Manifold and DNA isolation/purification columns (e.g., NucleoSpin™ NucleoFast™ or NucleoSnap™ kits) |
| For Use With (Application) | For controlling of vacuum |
| Quantity | 1 Ea. |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?