missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleostemin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15060-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.Specifications
Nucleostemin | |
Polyclonal | |
Unconjugated | |
GNL3 | |
C77032, E2 induced gene 3 protein, E2IG3, GNL3, NNP47, Novel nucleolar protein 47, NS, Nucleostemin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26354 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BVP2 | |
GNL3 | |
Nucleostemin Fusion Protein Ag7056 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |