Learn More
Macherey-Nagel™ NucleoSpin™ Soil, Mini kit for DNA from soil
Product Code. 11962402
Change view
Click to view available options
No. of Reactions:
10 Preps
250
50 Preps
Unit Size:
10 reactions
250 reactions
50 reactions
Product Code. No. of Reactions unitSize
11962402 50 Preps 50 reactions
12979884 10 Preps 10 reactions
12718562 250 250 reactions
3 options
Product Code. 11962402
Brand: Macherey-Nagel™ 740780.50
Isolate DNA from soil. Macherey Nagel™ NucleoSpin™ enables soil Isolation of total DNA from diverse soil types
Features
  • Two alternative lysis buffers and a special additive (Enhancer SX) for optimal processing of various soil samples – high yield and high purity
  • NucleoSpin™ Bead Tubes with ceramic beads for most efficient cell disruption of Gram-positive and Gram-negative bacteria, archaea, yeast, fungi, and algae in soil, sludge,or sediment
  • NucleoSpin™ Inhibitor Removal Column to eliminate all PCR inhibitors, such as humic substances – DNA is ready-to-use for PCR without dilution
Applications
  • Total DNA from microorganisms in soil and sediment
  • Quantify and detect microorganisms from environmental samples
  • Typical downstream applications: PCR, real-time PCR, Southern blotting, microarray technology
Specifications
Format Mini Spin Column
Isolation Technology Silica Membrane Technology
For Use With (Application) For Total DNA From Microorganisms in Soil and Sediment
No. of Reactions 50 Preps
Sample Volume <500 mg
Test Time 90min.
Yield 2 to 10μg
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate; guanidine hydrochloride, 2-propanol