Learn More
Macherey-Nagel™ NucleoSpin Dx RNA Blood, IVD Kit
Product Code. 18031091
Change view
Click to view available options
Quantity:
50 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18031091 50 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18031091 Supplier Macherey-Nagel™ Supplier No. 740201.50
  • ONLINE EXCLUSIVE PRICE
    6445.00 NOK
    6090.44 NOK / Pack of 1
    Save 354.56 NOK
    5% Off
 Restricted Item
In Stock
NucleoSpin Dx RNA Blood (IVD) for RNA isolation S-Monovette RNA Exact Isolation of RNA from the SARSTEDT's S-Monovette RNA Exact.
  • The NucleoSpin Dx RNA Blood kit is a IVD mini spin kit for the isolation of human RNA from whole blood collected in SARSTEDT's S-Monovette™ RNA Exact for the subsequent in-vitro diagnostic analysis (compliant with IVDR 2017/746).
  • The product provides purified human RNA to be used for subsequent down-stream analysis such as RT-PCR, qRT PCR, or RNA-Seq to obtain information about the RNA expression level in the sample.
  • The product is used by professional users in diagnostic laboratories.
Specifications
Content And Storage 50 Preps, 15 to 25 °C / 59 to 77 °F
Description Mini prep
Isolation Technology Silica Membrane Technology
Elution Volume 60 μL or 40 μL
Sample Type Whole Blood (Specific)
Efficiency Binding capacity: 200 μg
For Use With (Application) Isolation of RNA
No. of Reactions 50 Preps
Product Line NucleoSpin
Product Type IVD Kit
Purity 1.6 to 2.2 (typically 1.9 to 2.1)
Quantity 50 Preps
Sample Size 1.2 mL solution of stabilized blood from S-Monovette™ RNA Exact blood collection tubes (SARSTEDT REF 01.2048.001)
Target RNA
Test Time 55 min./6 Preps
Yield >1 μg (0.7 to 4.2 μg) per preparation from blood from healthy subjects