Learn More
Macherey-Nagel™ Nucleosil™ C18 EH PAH Analysis HPLC Column, 5 μm, 3 mm x 150 mm
Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 3 mm |
| For Use With | HPLC |
| Length (Metric) | 150 mm |
| Max. Pressure | 5800 psi (400 bar) |
| Particle Size | 5 μm |
| pH | 2 to 8 |
| Pore Size | 100 Å |
| Quantity | 1 Ea. |
| Surface Area | 350 m²/g |
| Show More |