Learn More
Macherey-Nagel™ Nucleosil™ 100-5 C18 720141 Model EC HPLC Column
Product Code. 12452660 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric):
4 mm
4.6 mm
Unit Size:
Each
Product Code. 12452660
Brand: Macherey-Nagel™ 720141.40
Only null left
Add to basket
Add to basket
Column Format: Analytical Column; Carbon Load: 15%; Pore Size: 100 Å; Column Phase: Reversed Phase
Specifications
Column Format Analytical Column
Crosslinking Octadecyl
Diameter (Metric) 4 mm
For Use With HPLC
Length (Metric) 100 mm
Max. Pressure 5800 psi (400 bar)
Mobile Phase Compatibility Acetonitrile/water
Particle Size 5 μm
pH 2 to 8
Pore Size 100 Å
Quantity 1 Ea.
Surface Area 350 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 15%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type EC Column