Learn More
Cytiva Nucleon™ PhytoPure™ Genomic DNA Extraction Kits
Product Code. 10762037 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mg
1000 mg
Unit Size:
50 reactions
Product Code. Quantity unitSize
10762037 100 mg 50 reactions
10410875 1000 mg 50 reactions
2 options
Product Code. 10762037
Brand: Cytiva RPN8510
Includes
Reagents 1 and 2 and PhytoPure Resin
Nucleon PhytoPure DNA Extraction kits use a phytopure resin to bind plant polysaccharides ensuring high yields of high-quality plant genomic DNA in only one hour
  • Rapid protocol does not require phenol or CTAB
  • Two kit sizes provide economical scale-up using the same phenol-free protocol
  • Effective removal of polysaccharides minimises inhibition of downstream applications
TRUSTED_SUSTAINABILITY
Specifications
Description Quality, length, and purity of a phenol-chloroform extraction without phenol. Nucleon PhytoPure enables rapid chloroform extraction of high-quality, high molecular weight genomic DNA from plant and fungal samples with efficient removal of protein as well
Sample Type Barley (Fresh, Frozen or Freeze Dried), Maize (Fresh, Frozen or Freeze Dried), Rye (Fresh, Frozen or Freeze Dried), Wheat (Fresh, Frozen or Freeze Dried)
For Use With (Application) Successfully extracts DNA from many plant species, starting with fresh, frozen, or freeze dried material
Includes Reagents 1 and 2 and PhytoPure™ Resin
Product Type illustra Nucleon PhytoPure