Learn More
Macherey-Nagel™ Nucleodur™ Phenyl/Hexyl Reversed Phase HPLC Column, 1.8 μm, 2 mm x 100 mm
Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
100 mm
Unit Size:
Each
Specifications
Specifications
| Column Format | Analytical Column |
| Crosslinking | Phenyl-hexyl |
| Diameter (Metric) | 2 mm |
| For Use With | For LC/MS and HPLC |
| Functionality/Interaction Type | p-p, Hydrophobic interactions |
| Length (Metric) | 100 mm |
| Max. Pressure | 13050 psi (900 bar) |
| Particle Shape | Spherical |
| Particle Size | 1.8 μm |
| pH | 1 to 10 |
| Show More |