Learn More
Macherey-Nagel™ Nucleodur™ C8 Gravity EC Standard Column
Product Code. 12458497 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
50 mm
125 mm
150 mm
Diameter (Metric):
2 mm
3 mm
4 mm
4.6 mm
Unit Size:
Each
Product Code. 12458497
Brand: Macherey-Nagel™ 760751.20
Only null left
Add to basket
Add to basket
Column Format: Analytical Column; Carbon Load: 11%; Pore Size: 110 Å; Column Phase: Reversed Phase
Specifications
Column Format Analytical Column
Diameter (Metric) 2 mm
Length (Metric) 125 mm
Max. Pressure 8700 psi (600 bar)
Mobile Phase Compatibility Acetonitrile/water
Particle Size 5 μm
pH 1 to 11
Pore Size 110 Å
Surface Area 340 m²/g
Temperature 60°C
USP Type L7
Stationary Phase C8 Gravity
Packing Material Silica
Carbon Load 11%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type EC Standard Column