Learn More
Macherey-Nagel™ Nucleodur™ C18 Isis HPLC Column
Product Code. p-8012214 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
125 mm
150 mm
250 mm
Diameter (Metric):
2 mm
3 mm
4 mm
4.6 mm
Particle Size:
1.8 μm
3 μm
5 μm
Carbon Load:
11%
20%
Unit Size:
Each
Product Code. 12757830
Brand: Macherey-Nagel™ 760409.20
Only null left
Specifications
Column Format Analytical Column
Crosslinking Polymeric Octadecyl
Diameter (Metric) 2 mm
For Use With For LC/MS and HPLC
Hydrophobicity No
Length (Metric) 150 mm
Max. Pressure 13050 psi (900 bar)
Particle Shape Spherical
Particle Size 1.8 μm
pH 1 to 10
Pore Size 110 Å
Quantity 1 Ea.
Solvents Acetonitrile/water
Surface Area 340 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18 Isis
Packing Material Silica
Carbon Load 20%
Column Type Steric Selectivity
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Type EC Column
Technique HPLC, LC/MS