Learn More
Macherey-Nagel™ Nucleodur™ C18 Isis HPLC Column
Product Code. 12428437 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
125 mm
150 mm
250 mm
Diameter (Metric):
2 mm
3 mm
4 mm
4.6 mm
Particle Size:
1.8 μm
3 μm
5 μm
Carbon Load:
11%
20%
Unit Size:
Each
Product Code. 12428437
Brand: Macherey-Nagel™ 760404.46
Only null left
Specifications
Column Chemistry Polymeric octadecyl
Column Format Analytical Column
Crosslinking Polymeric octadecyl
Diameter (Metric) 4.6 mm
Flow Rate 1 mL/min
For Use With For Steroids, (o,p,m-) Substituted Aromatics, Fat-Soluble Vitamins Application
Length (Metric) 250 mm
Max. Pressure 8700 psi (600 bar)
Mobile Phase Compatibility Acetonitrile/water
Particle Shape Spherical
Particle Size 3 μm
pH 1 to 10
Pore Size 110 Å
Pore Volume 0.9 mL/g
Quantity 1 Ea.
Solvents Acetonitrile/water
Surface Area 340 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18 Isis
Packing Material Silica
Carbon Load 20%
Column Type Steric Selectivity
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Type EC Column