missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra Midi Plus EF, Midi kit for endotoxin-free plasmid DNA
Click to view available options
No. of Reactions:
2
10
50
Unit Size:
0.1kg
2 reactions
50 preps
Description
Features
- Endotoxin-free
- Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Optimized column design – Midi prep in 45 min.
- Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
- Improved silica material – typical yields of 400μg (Midi)
- NucleoBond™ Xtra EF kits contain NucleoBond™ Finalizers to speed up DNA precipitation
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Specifications
| Buffer | Yes |
| Format | Midi Gravity-flow Column |
| Isolation Technology | Anion Exchange Chromatography Technology |
| No. of Reactions | 2 |
| RNase | Yes |
| Sample Volume | <200 mL (high copy), <400 mL (low copy) |
| Size | <300 kbp |
| Yield | 400μg |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : 2-propanol/ethanol (diluted < 20 %)/sodium hydroxide solution (diluted < 2 %), dodecyl sulfate, sodi
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction