Learn More
Macherey-Nagel™ NucleoBond™ PC EF, kit for endotoxin-free plasmid DNA
Product Code. p-8015713
Change view
Click to view available options
No. of Reactions:
5
10
Yield:
100–500μg
2,000–10,000μg
500–2,000μg
Sample Volume:
30 - 150mL
150 - 500mL
500 - 2000mL
Test Time:
100 min./2 preps
150 min./2 preps
180 min./2 preps
Unit Size:
10 reactions
5 preps
5 reactions
Product Code. No. of Reactions Yield Sample Volume Test Time unitSize
12863931 10 100–500μg 30 - 150mL 100 min./2 preps 10 reactions
12860661 5 500–2,000μg 150 - 500mL 150 min./2 preps 5 reactions
12873971 5 2,000–10,000μg 500 - 2000mL 180 min./2 preps 5 preps
3 options
Product Code. 12863931
Brand: Macherey-Nagel™ 740550
Removes endotoxins via an additional washing step, without ice incubation. Macherey-Nagel™ NucleoBond™ PC EF Kit is a classic anion exchanger for purification of endotoxin-free plasmid DNA. The kit includes NucleoBond™ AX Columns, all buffers, RNase A and NucleoBond™ Filters.
An important prerequisite for transfections and gene therapy applications is endotoxin-free plasmid DNA. The patented procedure of NucleoBond™ PC EF reduces endotoxins to a very low level using the same working procedure as the standard protocol. In comparison to competitive kits, there is no need for a time-consuming incubation step to remove the endotoxins.
Features
  • Fast and easy clarification by NucleoBond™ Folded Filters—No centrifugation required, no shearing forces
  • Proven anion-exchange technology—Yields transfection-grade high-copy plasmid DNA from E. coli
  • NucleoBond™ Columns contain NucleoBond™ AX Silica Resin packed between inert filters
  • Polypropylene columns are resistant to organic solvents and are free of DNase and RNase
  • AX Resin can be used over a wide pH range and can remain in contact with buffers for up to three hours
Specifications
  • DNA with less than 0.1 EU/μg DNA
  • Purity of plasmid DNA: A260/A280 = 1.80–1.95
  • Recovery of plasmid DNA >90%
  • Structural integrity >90%
  • Yield between 500μg up to 10mg, depending on kit used
Specifications
Concentration Endotoxin level: <0.1 EU/μg
Format Maxi Gravity-flow Column
Isolation Technology Anion Exchange Chromatography Technology
Efficiency Recovery of plasmid DNA: >90%
For Use With (Application) High-copy plasmids from E. coli
Includes NucleoBond AX 500 Columns, buffers, RNase A, NucleoBond Folded Filters XL
No. of Reactions 10
Product Type NucleoBond™ PC 500 EF
Quantity 1 Kit
Purity A260/A280: 1.80–1.95
RNase Yes
Sample Volume 30 - 150mL
Size <300 kbp
Test Time 100 min./2 preps
Yield 100–500μg
Show More Show Less
compliance-icons compliance-icons compliance-icons
Hazard Category
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1