missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSE gamma Mouse, Clone: ENO2/1375, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2026-MSM1-P0
Additional Details : Weight : 0.01000kg
Specifications
NSE gamma | |
Cytoplasmic | |
ENO2/1375 | |
Unconjugated | |
NP_001966 | |
Mouse | |
50kDa | |
Protein A/G Purified | |
RUO | |
2026 | |
IgG2b |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
ENO2 | |
A synthetic peptide corresponding to aa416-433 of human NSE gamma | |
HepG2, SH-SY-5Y, HeLa or Y79 cells. Pancreas, Cerebellum or Pheochromocytoma. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |