missing translation for 'onlineSavingsMsg'
Learn More

NSD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11345-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888353

  • 1281.92 NOK / 20µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody recognize the long isoform (160-170 kDa) and short isoform (68-72 kDa).

This gene is related to the Wolf-Hirschhorn syndrome candidate-1 gene and encodes a protein with PWWP (proline-tryptophan-tryptophan-proline) domains. The function of the protein has not been determined. Two alternatively spliced variants have been described.
Specifications

Specifications

NSD3
Polyclonal
Unconjugated
NSD3
NSD3, Protein whistle, WHSC1 like protein 1, WHSC1L1
Rabbit
Antigen Affinity Chromatography
RUO
234135, 290831, 54904
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6P2L6, Q9BZ95
Nsd3
NSD3 Fusion Protein Ag1894
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers