Learn More
NSC319726, MedChemExpress
Product Code. 16423577
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16423577 50 mg 50mg
16460028 5 mg 5mg
16407558 10 mg 10mg
3 options
Product Code. 16423577
Brand: Medchem Express HY18634
MedChemExpress NSC319726 (ZMC1) is a mutant p53R175 reactivator; inhibits growth of fibroblasts expressing the p53R175 mutation (IC50 = 8 nM); shows no inhibition for p53 wild-type cells.
NSC319726 (ZMC1) is a mutant p53R175 reactivator
Specifications
Chemical Name or Material NSC319726
CAS 71555-25-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H14N4S
Quantity 50 mg
Synonym ZMC1
Solubility Information DMSO : 31.25 mg/mL (133.36 mM; Need ultrasonic)
SMILES C/C(C1=NC=CC=C1)=N\NC(N2CCC2)=S
Molecular Weight (g/mol) 234.32
Formula Weight 234.32
Percent Purity 98.06%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death