Learn More
Abnova™ NRGN (Human) Recombinant Protein
Product Code. p-7164519
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16118501
Brand: Abnova™ H00004900P01.10ug
Human NRGN full-length ORF ( AAH02835, 1 a.a. - 78 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MDCCTENACSKPDDDILDIPLDDPGANAAAAKIQASFRGHMARKKIKSGERGRKGPGPGGPGGAGVARGGAGGGPSGD
Specifications
Accession Number AAH02835
Gene ID (Entrez) 4900
Name neurogranin (protein kinase C substrate, RC3)
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias RC3, hng
Gene Symbol NRGN
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer