Learn More
NRC-2694, MedChemExpress
Product Code. 16487958
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
Product Code. 16487958
Brand: Medchem Express HY19909
MedChemExpress NRC-2694 is an epidermal growth factor receptor (EGFR) antagonist with anti-cancer and anti-proliferative properties.
NRC-2694 is an epidermal growth factor receptor (EGFR) antagonist with anti-cancer and anti-proliferative properties.
Specifications
Chemical Name or Material NRC-2694
CAS 936446-61-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H26N4O3
Quantity 10 mg
Solubility Information DMSO : 250 mg/mL (597.39 mM; Need ultrasonic)
SMILES COC1=CC2=NC=NC(NC3=CC=CC(C#C)=C3)=C2C=C1OCCCN4CCOCC4
Molecular Weight (g/mol) 418.49
Formula Weight 418.49
Percent Purity 98.4%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.