missing translation for 'onlineSavingsMsg'
Learn More

NRAS-Specific Rabbit anti-Human, Rat, Polyclonal, Proteintech

Product Code. p-7225914 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17669606 20 μL 20µL
17659606 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 17669606

Brand: Proteintech 182961AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NRAS-Specific Polyclonal antibody apecifically detetects NRAS-Specific antigen in Human, Rat Samples. It is validated for Western Blot, ELISA.

Specifications

Antigen NRAS-Specific
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 350 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC005219
Gene Symbols NRAS
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 4893
Target Species Human, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.