Learn More
NR4A2 Mouse anti-Human, Unconjugated, Clone: 1H5B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66878-1-IG-150UL
This item is not returnable.
View return policy
Description
The NR4A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NR4A2. This antibody recognizes human, mouse antigen. The NR4A2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NR4A2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot | |
| 1H5B4 | |
| Unconjugated | |
| 150 μL | |
| 4929 | |
| IgG1 |